Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576315.2 | XP_005576372.1 | 1 - 2 | 384 | 99 | |
XM_005576316.2 | XP_005576373.1 | 1 - 2 | 384 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |