Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581844.2 | XP_005581901.1 | 4 - 5 | 1110 | 89 | |
XM_015437237.1 | XP_015292723.1 | 4 - 5 | 1156 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |