Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005545025.2 | XP_005545082.1 | 6 - 7 | 701 | 99 | |
XM_005545026.2 | XP_005545083.1 | 6 - 7 | 3829 | 99 | ||
XM_005545027.2 | XP_005545084.1 | 6 - 7 | 587 | 99 | ||
XM_015443766.1 | XP_015299252.1 | 5 - 6 | 1010 | 99 | ||
XM_015443767.1 | XP_015299253.1 | 4 - 5 | 905 | 99 | ||
XM_015443769.1 | XP_015299255.1 | 6 - 7 | 3833 | 99 | ||
XM_015443770.1 | XP_015299256.1 | 6 - 7 | 646 | 99 | ||
XM_015443774.1 | XP_015299260.1 | 6 - 7 | 529 | 99 | ||
XM_015443777.1 | XP_015299263.1 | 6 - 7 | 529 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |