Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005564688.2 | XP_005564745.1 | 2 - 3 | 372 | 143 | |
XM_005564689.2 | XP_005564746.1 | 2 - 3 | 274 | 143 | ||
XM_005564690.2 | XP_005564747.1 | 2 - 3 | 259 | 143 | ||
XM_005564692.1 | XP_005564749.1 | 2 - 3 | 183 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |