Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005542805.2 | XP_005542862.1 | 1 - 2 | 373 | 60 | |
GenBank mRNA | AB179271.1 | - | 1 - 2 | 105 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |