Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587696.2 | XP_005587753.1 | 3 - 4 | 942 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |