Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284703.1 | NP_001271632.1 | 2 - 3 | 505 | 71 | |
XM_005563233.2 | XP_005563290.1 | 2 - 3 | 319 | 71 | ||
GenBank mRNA | AB179209.1 | - | 2 - 3 | 505 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |