Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578714.2 | XP_005578771.1 | 3 - 4 | 539 | 86 | |
XM_005578715.2 | XP_005578772.1 | 3 - 4 | 539 | 86 | ||
XM_005578716.2 | XP_005578773.1 | 5 - 6 | 667 | 86 | ||
XM_005578717.2 | XP_005578774.1 | 5 - 6 | 611 | 86 | ||
XM_005578718.2 | XP_005578775.1 | 4 - 5 | 526 | 86 | ||
XM_005578719.2 | XP_005578776.1 | 3 - 4 | 261 | 86 | ||
XM_015435176.1 | XP_015290662.1 | 3 - 4 | 539 | 86 | ||
XM_015435177.1 | XP_015290663.1 | 5 - 6 | 1264 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |