Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLPH3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283571.1 | NP_001270500.1 | 3 - 4 | 518 | 104 | |
XM_005541976.2 | XP_005542033.1 | 4 - 5 | 646 | 104 | ||
GenBank mRNA | AB171843.1 | - | 3 - 4 | 518 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |