Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283593.1 | NP_001270522.1 | 1 - 2 | 72 | 67 | |
GenBank mRNA | AB083330.2 | - | 1 - 2 | 67 | ||
AB169529.1 | - | 1 - 2 | 93 | 67 | ||
AB613820.1 | - | 1 - 2 | 72 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |