Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285098.1 | NP_001272027.1 | 4 - 5 | 391 | 84 | |
XM_015442172.1 | XP_015297658.1 | 4 - 5 | 437 | 84 | ||
XM_015442173.1 | XP_015297659.1 | 4 - 5 | 437 | 84 | ||
XM_015442174.1 | XP_015297660.1 | 5 - 6 | 818 | 84 | ||
XM_015442175.1 | XP_015297661.1 | 5 - 6 | 516 | 84 | ||
XM_015442177.1 | XP_015297663.1 | 5 - 6 | 609 | 84 | ||
XM_015442178.1 | XP_015297664.1 | 4 - 5 | 703 | 84 | ||
XM_015442179.1 | XP_015297665.1 | 4 - 5 | 482 | 84 | ||
XM_015442180.1 | XP_015297666.1 | 5 - 6 | 567 | 84 | ||
GenBank mRNA | AB125191.1 | - | 4 - 5 | 391 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |