Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005580510.2 | XP_005580567.1 | 7 - 8 | 1395 | 112 | |
XM_005580512.2 | XP_005580569.1 | 5 - 6 | 829 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |