Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005558398.2 | XP_005558455.1 | 5 - 6 | 973 | 89 | |
XM_015452197.1 | XP_015307683.1 | 5 - 6 | 662 | 89 | ||
XM_015452198.1 | XP_015307684.1 | 5 - 6 | 1002 | 89 | ||
XM_015452199.1 | XP_015307685.1 | 5 - 6 | 878 | 89 | ||
XM_015452200.1 | XP_015307686.1 | 4 - 5 | 462 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |