Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285062.1 | NP_001271991.1 | 2 - 3 | 366 | 64 | |
XM_015435348.1 | XP_015290834.1 | 4 - 5 | 683 | 64 | ||
XM_015435349.1 | XP_015290835.1 | 4 - 5 | 619 | 64 | ||
XM_015435350.1 | XP_015290836.1 | 3 - 4 | 542 | 64 | ||
GenBank mRNA | AB125159.1 | - | 2 - 3 | 366 | 64 | |
AB170174.1 | - | 2 - 3 | 366 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |