Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005540297.2 | XP_005540354.1 | 4 - 5 | 784 | 76 | |
XM_005540298.2 | XP_005540355.1 | 4 - 5 | 784 | 76 | ||
XM_005540299.2 | XP_005540356.1 | 4 - 5 | 784 | 76 | ||
XM_015447396.1 | XP_015302882.1 | 4 - 5 | 784 | 76 | ||
XM_015447397.1 | XP_015302883.1 | 4 - 5 | 784 | 76 | ||
XM_015447399.1 | XP_015302885.1 | 4 - 5 | 784 | 76 | ||
GenBank mRNA | JX277090.1 | - | 4 - 5 | 496 | 76 | |
JX277091.1 | - | 4 - 5 | 496 | 76 | ||
JX277094.1 | - | 4 - 5 | 496 | 76 | ||
JX277095.1 | - | 4 - 5 | 496 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Inventoried Probe spans exons |