Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589957.2 | XP_005590014.1 | 4 - 5 | 1330 | 71 | |
XM_005589958.2 | XP_005590015.1 | 4 - 5 | 1349 | 71 | ||
XM_015441178.1 | XP_015296664.1 | 4 - 5 | 3721 | 71 | ||
XM_015441180.1 | XP_015296666.1 | 4 - 5 | 781 | 71 | ||
XM_015441181.1 | XP_015296667.1 | 4 - 5 | 734 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |