Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102144847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544596.2 | XP_005544653.1 | 5 - 6 | 1229 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |