Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102118724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005594220.2 | XP_005594277.1 | 2 - 3 | 510 | 139 | |
XM_005594222.1 | XP_005594279.1 | 2 - 3 | 258 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |