Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005594215.2 | XP_005594272.1 | 3 | 897 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |