Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005580331.2 | XP_005580388.1 | 1 - 2 | 877 | 80 | |
XM_005580332.2 | XP_005580389.1 | 1 - 2 | 877 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |