Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16H17orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584763.2 | XP_005584820.1 | 1 - 2 | 199 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |