Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283910.1 | NP_001270839.1 | 5 - 6 | 473 | 109 | |
XM_015435356.1 | XP_015290842.1 | 5 - 6 | 571 | 109 | ||
XM_015435357.1 | XP_015290843.1 | 5 - 6 | 571 | 109 | ||
XM_015435358.1 | XP_015290844.1 | 4 - 5 | 528 | 109 | ||
GenBank mRNA | AB073992.1 | - | 5 - 6 | 473 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Inventoried Probe spans exons |