Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581865.2 | XP_005581922.1 | 8 - 9 | 1168 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |