Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593073.2 | XP_005593130.1 | 2 - 3 | 3548 | 104 | |
XM_005593074.2 | XP_005593131.1 | 2 - 3 | 3551 | 104 | ||
XM_005593075.2 | XP_005593132.1 | 3 - 4 | 2319 | 104 | ||
XM_005593076.2 | XP_005593133.1 | 4 - 5 | 519 | 104 | ||
XM_005593077.2 | XP_005593134.1 | 2 - 3 | 3551 | 104 | ||
XM_005593078.2 | XP_005593135.1 | 3 - 4 | 2316 | 104 | ||
XM_005593079.2 | XP_005593136.1 | 4 - 5 | 538 | 104 | ||
XM_005593082.2 | XP_005593139.1 | 3 - 4 | 368 | 104 | ||
XM_015443358.1 | XP_015298844.1 | 3 - 4 | 1749 | 104 | ||
XM_015443359.1 | XP_015298845.1 | 3 - 4 | 2316 | 104 | ||
XM_015443360.1 | XP_015298846.1 | 4 - 5 | 536 | 104 | ||
XM_015443361.1 | XP_015298847.1 | 3 - 4 | 365 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |