Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579110.2 | XP_005579167.1 | 4 - 5 | 1241 | 141 | |
XM_005579111.1 | XP_005579168.1 | 5 - 6 | 1290 | 141 | ||
XM_015435503.1 | XP_015290989.1 | 5 - 6 | 1080 | 141 | ||
XM_015435504.1 | XP_015290990.1 | 4 - 5 | 1002 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |