Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588011.2 | XP_005588068.1 | 4 - 5 | 755 | 124 | |
XM_015440534.1 | XP_015296020.1 | 4 - 5 | 747 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |