Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553103.2 | XP_005553160.1 | 1 - 2 | 269 | 84 | |
XM_005553104.2 | XP_005553161.1 | 1 - 2 | 269 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |