Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CANT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585148.2 | XP_005585205.1 | 3 - 4 | 974 | 86 | |
XM_005585149.2 | XP_005585206.1 | 4 - 5 | 1076 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |