Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIVEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551999.2 | XP_005552056.1 | 7 - 8 | 17696 | 73 | |
XM_015448728.1 | XP_015304214.1 | 8 - 9 | 17764 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |