Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544274.2 | XP_005544331.1 | 3 - 4 | 497 | 92 | |
XM_005544276.2 | XP_005544333.1 | 3 - 4 | 1769 | 92 | ||
XM_015439968.1 | XP_015295454.1 | 4 - 5 | 588 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |