Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRICKLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553036.2 | XP_005553093.1 | 5 - 6 | 645 | 129 | |
XM_005553037.2 | XP_005553094.1 | 5 - 6 | 503 | 129 | ||
XM_015449409.1 | XP_015304895.1 | 4 - 5 | 1065 | 129 | ||
XM_015449410.1 | XP_015304896.1 | 5 - 6 | 645 | 129 | ||
XM_015449411.1 | XP_015304897.1 | 4 - 5 | 1065 | 129 | ||
XM_015449412.1 | XP_015304898.1 | 5 - 6 | 1129 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |