Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXPH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015435671.1 | XP_015291157.1 | 1 - 2 | 211 | 98 | |
XM_015435672.1 | XP_015291158.1 | 1 - 2 | 334 | 98 | ||
XM_015435673.1 | XP_015291159.1 | 1 - 2 | 334 | 98 | ||
XM_015435674.1 | XP_015291160.1 | 1 - 2 | 334 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |