Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555058.1 | XP_005555115.1 | 2 - 3 | 1045 | 92 | |
XM_015450394.1 | XP_015305880.1 | 3 - 4 | 888 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |