Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283203.1 | NP_001270132.1 | 2 - 3 | 561 | 131 | |
GenBank mRNA | AB063100.1 | - | 2 - 3 | 561 | 131 | |
AF461182.1 | - | 2 - 3 | 426 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |