Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588533.2 | XP_005588590.1 | 2 - 3 | 875 | 68 | |
XM_015440875.1 | XP_015296361.1 | 1 - 2 | 313 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |