Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553498.2 | XP_005553555.1 | 3 - 4 | 776 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |