Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593740.2 | XP_005593797.1 | 1 - 2 | 435 | 116 | |
XM_005593741.2 | XP_005593798.1 | 1 - 2 | 435 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |