Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RINL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015441435.1 | XP_015296921.1 | 7 - 8 | 963 | 104 | |
XM_015441436.1 | XP_015296922.1 | 7 - 8 | 963 | 104 | ||
XM_015441438.1 | XP_015296924.1 | 7 - 8 | 652 | 104 | ||
XM_015441439.1 | XP_015296925.1 | 7 - 8 | 580 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |