Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566494.2 | XP_005566551.1 | 6 - 7 | 1708 | 131 | |
XM_005566495.2 | XP_005566552.1 | 6 - 7 | 1977 | 131 | ||
XM_005566496.2 | XP_005566553.1 | 6 - 7 | 818 | 131 | ||
XM_005566499.2 | XP_005566556.1 | 6 - 7 | 1972 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |