Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571700.2 | XP_005571757.1 | 2 - 3 | 272 | 126 | |
XM_015431302.1 | XP_015286788.1 | 2 - 3 | 275 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |