Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015452016.1 | XP_015307502.1 | 1 | 4365 | 186 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Both primers and probe map within a single exon |