Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102120484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570925.2 | XP_005570982.1 | 1 | 1461 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Both primers and probe map within a single exon |