Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549929.2 | XP_005549986.1 | 2 - 3 | 1447 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |