Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287678.1 | NP_001274607.1 | 2 | 1457 | 63 | |
GenBank mRNA | AB446510.1 | - | 1 | 848 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Inventoried Both primers and probe map within a single exon |