Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583572.2 | XP_005583629.1 | 1 - 2 | 694 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |