Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015452646.1 | XP_015308132.1 | 2 | 1615 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |