Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557673.2 | XP_005557730.1 | 2 - 3 | 2170 | 97 | |
GenBank mRNA | AB171287.1 | - | 2 - 3 | 810 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |