Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015430083.1 | XP_015285569.1 | 2 - 3 | 538 | 65 | |
XM_015430084.1 | XP_015285570.1 | 2 - 3 | 535 | 65 | ||
XM_015430085.1 | XP_015285571.1 | 2 - 3 | 533 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |