Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587940.2 | XP_005587997.1 | 3 - 4 | 1897 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |