Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015430981.1 | XP_015286467.1 | 1 - 2 | 6864 | 114 | |
XM_015430982.1 | XP_015286468.1 | 2 - 3 | 6749 | 114 | ||
XM_015430983.1 | XP_015286469.1 | 2 - 3 | 6719 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |